Learn More
MSC Value Collection Tabletop Thermal Sealer, 12 in Seal, 1 ea
Catalog No. 5000781615
Encompass
This tabletop thermal sealer from Value Collection is designed for efficient sealing of polybags and other materials. It features an adjustable timer to achieve perfect seals across various film thicknesses.
  • Adjustable timer for versatile sealing of poly thicknesses
  • Maximum 12 in seal size
Catalog No. 50-007-81615 Supplier MSC Supplier No. 46006292
This tabletop thermal sealer from Value Collection is designed for efficient sealing of polybags and other materials. It features an adjustable timer to achieve perfect seals across various film thicknesses.
  • Adjustable timer for versatile sealing of poly thicknesses
  • Maximum 12 in seal size