Learn More
MSC Value Collection Telescoping Gage Set, 5/16 to 6 in, 6 Pc
Catalog No. 5000516810
Encompass
This telescoping gage set is designed for precise measurement applications. It includes 6 pieces, made from hardened tool steel with a satin chrome finish, covering a measurement range of 5/16 to 6 inches.
  • Minimum measurement of 5/16 "
  • Maximum measurement of 6 inches.
  • Includes 6 pieces.
  • Constructed from hardened tool steel.
  • Features a satin chrome finish.
Catalog No. 50-005-16810 Supplier MSC Supplier No. 06456016
This telescoping gage set is designed for precise measurement applications. It includes 6 pieces, made from hardened tool steel with a satin chrome finish, covering a measurement range of 5/16 to 6 inches.
  • Minimum measurement of 5/16 "
  • Maximum measurement of 6 inches.
  • Includes 6 pieces.
  • Constructed from hardened tool steel.
  • Features a satin chrome finish.