Learn More
MSC Value Collection Twisted Rope, Polypropylene, 5/8 in, 1200 ft
Catalog No. 5000532759
Encompass
This polypropylene twisted rope is available in 1 ft. increments, with a total coil length of 1200 ft. It features a 5/8 in diameter and a capacity of 697 lb. The rope is made of polypropylene and has a twisted construction.
  • Quality products at a great value to support your business needs.
  • Available in 1 ft. increments for customizable lengths.
  • Item is cut to your specifications.
  • Not returnable due to its custom-cut nature.
  • Special handling applies to cut-to-length items, and shipping dates are not guaranteed.
Catalog No. 50-005-32759 Supplier MSC Supplier No. 45900818
This polypropylene twisted rope is available in 1 ft. increments, with a total coil length of 1200 ft. It features a 5/8 in diameter and a capacity of 697 lb. The rope is made of polypropylene and has a twisted construction.
  • Quality products at a great value to support your business needs.
  • Available in 1 ft. increments for customizable lengths.
  • Item is cut to your specifications.
  • Not returnable due to its custom-cut nature.
  • Special handling applies to cut-to-length items, and shipping dates are not guaranteed.