Learn More
MSC Value Collection Twisted Seine Twine, #36 Twine Dia, Nylon, White
Catalog No. 5000532365
Encompass
This Twisted Seine Twine features a #36 twine diameter, constructed from durable nylon in a white color. It offers a breaking strength of 279 lb and provides 536 feet of twine per pound.
  • Quality product at a great value.
  • Durable with a 279 lb breaking strength.
  • Provides 536 feet of twine per pound for extended use.
Catalog No. 50-005-32365 Supplier MSC Supplier No. 45891058
This Twisted Seine Twine features a #36 twine diameter, constructed from durable nylon in a white color. It offers a breaking strength of 279 lb and provides 536 feet of twine per pound.
  • Quality product at a great value.
  • Durable with a 279 lb breaking strength.
  • Provides 536 feet of twine per pound for extended use.