Learn More
MSC Value Collection Twisted Seine Twine, Nylon, #72 Twine, 567 lb Breaking Strength
Catalog No. 5000528558
Encompass
This Twisted Seine Twine is constructed from durable nylon, designed for high-strength applications such as fishing nets and general utility. Featuring a #72 twine diameter, it offers a robust breaking strength of 567 lb for reliable performance.
  • Twisted seine twine design
  • Durable nylon construction
  • #72 twine diameter
  • High breaking strength of 567 lb
  • 262 feet per pound length
Catalog No. 50-005-28558 Supplier MSC Supplier No. 45891074
This Twisted Seine Twine is constructed from durable nylon, designed for high-strength applications such as fishing nets and general utility. Featuring a #72 twine diameter, it offers a robust breaking strength of 567 lb for reliable performance.
  • Twisted seine twine design
  • Durable nylon construction
  • #72 twine diameter
  • High breaking strength of 567 lb
  • 262 feet per pound length