Learn More
MSC Value Collection V-Block, 2-9/16 in Capacity, 90° V, Hardened Steel
Catalog No. 5000528444
Encompass
This V-block is constructed from hardened steel and designed for precision applications, featuring a maximum capacity of 2-9/16 inches and a 90° V angle. It offers high accuracy with a V parallelism and squareness of 0.0002 inch, ensuring reliable performance. This individual unit comes complete with a wooden case and a clamp for convenient storage and secure workpiece holding.
  • Constructed from hardened steel
  • Maximum capacity of 2-9/16 inches
  • V angle of 90 degrees
  • V parallelism of 0.0002 inch
  • Squareness of 0.0002 inch
  • Sold individually
  • Includes a wooden case and clamp
  • Non-magnetic
Catalog No. 50-005-28444 Supplier MSC Supplier No. 06493126
This V-block is constructed from hardened steel and designed for precision applications, featuring a maximum capacity of 2-9/16 inches and a 90° V angle. It offers high accuracy with a V parallelism and squareness of 0.0002 inch, ensuring reliable performance. This individual unit comes complete with a wooden case and a clamp for convenient storage and secure workpiece holding.
  • Constructed from hardened steel
  • Maximum capacity of 2-9/16 inches
  • V angle of 90 degrees
  • V parallelism of 0.0002 inch
  • Squareness of 0.0002 inch
  • Sold individually
  • Includes a wooden case and clamp
  • Non-magnetic