Learn More
MSC Value Collection V-Block Clamp, 1-1/4 Inch Max Capacity, Cast Iron, 90° V Angle
Catalog No. 5000527833
Encompass
This V-Block Clamp is designed for use with Series 030 V-Blocks, providing secure workholding. It features a 1-1/4 inch maximum capacity and is constructed from durable cast iron, offering reliable performance for various applications.
  • 90° V angle
  • Cast iron construction
  • Designed for use with Series 030 V-Block
Catalog No. 50-005-27833 Supplier MSC Supplier No. 06494306
This V-Block Clamp is designed for use with Series 030 V-Blocks, providing secure workholding. It features a 1-1/4 inch maximum capacity and is constructed from durable cast iron, offering reliable performance for various applications.
  • 90° V angle
  • Cast iron construction
  • Designed for use with Series 030 V-Block