Learn More
MSC Value Collection V Groove Caster, 6 in Wheel Diameter, 2 in Wheel Width, 1000 lb Capacity
Catalog No. 5000510560
Encompass
This V Groove Caster from Value Collection is a durable and reliable component designed for demanding industrial environments. It features a robust cold-forged frame, a plate mount style, and a metal wheel with a zinc finish, providing excellent performance for various applications.
  • Plate style mount
  • Roller swivel bearing type
  • 1000 lb load capacity
  • Cold forged frame fabrication
  • Metal wheel material
  • Zinc finish
  • Suitable for production machinery applications
  • Compatible with 4 x 4-1/2 top plates
Catalog No. 50-005-10560 Supplier MSC Supplier No. 66030065
This V Groove Caster from Value Collection is a durable and reliable component designed for demanding industrial environments. It features a robust cold-forged frame, a plate mount style, and a metal wheel with a zinc finish, providing excellent performance for various applications.
  • Plate style mount
  • Roller swivel bearing type
  • 1000 lb load capacity
  • Cold forged frame fabrication
  • Metal wheel material
  • Zinc finish
  • Suitable for production machinery applications
  • Compatible with 4 x 4-1/2 top plates