Learn More
MSC Value Collection Vernier Height Gage, 0 to 24 Inch Measurement, 0.001 Inch Graduation
Catalog No. 5000529362
Encompass
This vernier height gage is engineered for accurate and reliable measurements in various applications. Constructed from durable steel with a satin chrome finish, it offers a broad measurement range and fine graduation for precise readings.
  • Measures from 0 to 24 in
  • Provides a graduation of 0.001 in
  • Features a satin chrome finish for durability
  • Constructed from high-quality steel
  • Dimensions are 7 in long x 4.7 in wide
Catalog No. 50-005-29362 Supplier MSC Supplier No. 06431340
This vernier height gage is engineered for accurate and reliable measurements in various applications. Constructed from durable steel with a satin chrome finish, it offers a broad measurement range and fine graduation for precise readings.
  • Measures from 0 to 24 in
  • Provides a graduation of 0.001 in
  • Features a satin chrome finish for durability
  • Constructed from high-quality steel
  • Dimensions are 7 in long x 4.7 in wide