Learn More
MSC Value Collection Welded Alloy Chain, 9/32 inch, Grade 80, 3,500 Lb
Catalog No. 5000530356
Encompass
This 9/32 inch welded alloy chain is designed for heavy-duty applications, offering a robust solution for lifting and securing. With a Grade 80 rating and a substantial capacity of 3,500 Lb, it provides reliable performance in demanding environments. The chain features a cataphoresis finish for enhanced durability.
  • Designed for heavy-duty lifting and securing applications.
  • Provides robust and reliable performance.
  • Features a durable cataphoresis finish.
  • Offers a substantial working load capacity of 3,500 Lb.
  • Grade 80 chain for demanding environments.
  • 800 ft. total length.
Catalog No. 50-005-30356 Supplier MSC Supplier No. 66795360
This 9/32 inch welded alloy chain is designed for heavy-duty applications, offering a robust solution for lifting and securing. With a Grade 80 rating and a substantial capacity of 3,500 Lb, it provides reliable performance in demanding environments. The chain features a cataphoresis finish for enhanced durability.
  • Designed for heavy-duty lifting and securing applications.
  • Provides robust and reliable performance.
  • Features a durable cataphoresis finish.
  • Offers a substantial working load capacity of 3,500 Lb.
  • Grade 80 chain for demanding environments.
  • 800 ft. total length.