Learn More
MSC Value Collection Welded Proof Coil Chain, 1/2 in, 4,500 Lb Capacity, Grade 30, Zinc Plated
Catalog No. 5000514086
Encompass
The Value Collection 1/2 in welded proof coil chain is designed for various applications, offering a robust 4,500 Lb capacity. It features a durable Grade 30 construction with a protective zinc plated finish, ensuring reliability and longevity. This chain is available in 1 ft increments with a total coil length of 200 ft.
  • Trade size: 1/2 in
  • Zinc plated finish
  • 200 ft overall length
  • Grade 30 chain
  • 4,500 Lb working load limit
  • Suitable for cut-to-length requirements
  • Country of origin: China
Catalog No. 50-005-14086 Supplier MSC Supplier No. 66343757
The Value Collection 1/2 in welded proof coil chain is designed for various applications, offering a robust 4,500 Lb capacity. It features a durable Grade 30 construction with a protective zinc plated finish, ensuring reliability and longevity. This chain is available in 1 ft increments with a total coil length of 200 ft.
  • Trade size: 1/2 in
  • Zinc plated finish
  • 200 ft overall length
  • Grade 30 chain
  • 4,500 Lb working load limit
  • Suitable for cut-to-length requirements
  • Country of origin: China