Learn More
MSC Value Collection Welded Stainless Steel Chain, 9/32 in, 2,000 lb
Catalog No. 5000512521
Encompass
This welded stainless steel chain offers a 2,000 lb capacity, suitable for various industrial and commercial applications. It features a 9/32 inch diameter and a bright finish, providing durability and a clean appearance.
  • 2,000 lb capacity
  • 9/32 in diameter
  • Bright finish
  • Welded link type
  • 0.500 in inside width
  • 1.00 in inside length
  • 14 links per foot
Catalog No. 50-005-12521 Supplier MSC Supplier No. 60470556
This welded stainless steel chain offers a 2,000 lb capacity, suitable for various industrial and commercial applications. It features a 9/32 inch diameter and a bright finish, providing durability and a clean appearance.
  • 2,000 lb capacity
  • 9/32 in diameter
  • Bright finish
  • Welded link type
  • 0.500 in inside width
  • 1.00 in inside length
  • 14 links per foot