Learn More
MSC Value Collection Welded Standard Link Chain, 5/16, Grade 30, 62 ft, Zinc
Catalog No. 5000536714
Encompass
This welded standard link chain is designed for various applications, featuring a 5/16 trade size and Grade 30 strength. It comes in a 62 ft length and has a protective zinc finish for durability.
  • 5/16 trade size
  • Standard link type
  • Zinc finish
  • 62 ft overall length
  • Grade 30 chain
  • 0.46 in inside width
  • 1.24 in inside length
  • Country of origin: China
Catalog No. 50-005-36714 Supplier MSC Supplier No. 45659497
This welded standard link chain is designed for various applications, featuring a 5/16 trade size and Grade 30 strength. It comes in a 62 ft length and has a protective zinc finish for durability.
  • 5/16 trade size
  • Standard link type
  • Zinc finish
  • 62 ft overall length
  • Grade 30 chain
  • 0.46 in inside width
  • 1.24 in inside length
  • Country of origin: China