Learn More
MSC Value Collection Wire Rope, 1/2 in, Steel, 6x19 IWRC, 20700 Lb
Catalog No. 5000514899
Encompass
This wire rope features a 1/2 in diameter and a 6x19 IWRC strand type, designed for various industrial applications. It offers a robust breaking strength of 20,700 Lb, ensuring reliability and performance. This product is supplied in 1 ft increments, with a total coil length of 1,000 ft.
  • Has a breaking strength of 20,700 Lb.
  • Features a 6x19 IWRC strand type.
  • Available in 1 ft increments.
  • Comes in a 1,000 ft total coil length.
  • Has a bright, uncoated finish.
Catalog No. 50-005-14899 Supplier MSC Supplier No. 45701463
This wire rope features a 1/2 in diameter and a 6x19 IWRC strand type, designed for various industrial applications. It offers a robust breaking strength of 20,700 Lb, ensuring reliability and performance. This product is supplied in 1 ft increments, with a total coil length of 1,000 ft.
  • Has a breaking strength of 20,700 Lb.
  • Features a 6x19 IWRC strand type.
  • Available in 1 ft increments.
  • Comes in a 1,000 ft total coil length.
  • Has a bright, uncoated finish.