Learn More
MSC Value Collection Wire Rope, 1/8 in Dia, Galvanized Steel, 7x19 Strand Core
Catalog No. 5000511232
Encompass
This galvanized steel wire rope features a 1/8 inch diameter and a 7x19 strand core, providing a robust breaking strength of 2,000 Lb. It is suitable for various applications requiring strength and corrosion resistance.
  • Diameter: 1/8 in
  • Breaking strength: 2,000 Lb
  • Strand core: 7 x 19
  • Finish: Galvanized
  • Uncoated head coating
  • Length: 50 ft roll
Catalog No. 50-005-11232 Supplier MSC Supplier No. 45701661
This galvanized steel wire rope features a 1/8 inch diameter and a 7x19 strand core, providing a robust breaking strength of 2,000 Lb. It is suitable for various applications requiring strength and corrosion resistance.
  • Diameter: 1/8 in
  • Breaking strength: 2,000 Lb
  • Strand core: 7 x 19
  • Finish: Galvanized
  • Uncoated head coating
  • Length: 50 ft roll