Learn More
MSC Value Collection Wire Rope, 3/8 in Diameter, Steel, 6 x 19 IWRC
Catalog No. 5000511904
Encompass
This wire rope is a general-purpose steel wire rope with a 3/8 inch diameter and 6 x 19 IWRC construction, offering a breaking strength of 10,980 lb. It is designed to provide quality and value for various business needs and is available in 1-foot increments for cut-to-length requirements, with a total coil length of 1,000 feet.
  • 3/8 inch diameter
  • Steel material
  • 6 x 19 IWRC construction
  • 10,980 lb breaking strength
  • Bright finish
  • Uncoated head
  • Priced in 1-foot increments
  • Available as cut-to-length items
Catalog No. 50-005-11904 Supplier MSC Supplier No. 45701380
This wire rope is a general-purpose steel wire rope with a 3/8 inch diameter and 6 x 19 IWRC construction, offering a breaking strength of 10,980 lb. It is designed to provide quality and value for various business needs and is available in 1-foot increments for cut-to-length requirements, with a total coil length of 1,000 feet.
  • 3/8 inch diameter
  • Steel material
  • 6 x 19 IWRC construction
  • 10,980 lb breaking strength
  • Bright finish
  • Uncoated head
  • Priced in 1-foot increments
  • Available as cut-to-length items