Learn More
MSC Value Collection Wire Rope, 5/32 in Dia, Galvanized Steel, 2,800 Lb
Catalog No. 5000511905
Encompass
This galvanized steel wire rope features a 5/32 in diameter and a 7 x 19 strand core. It offers a 2,800 Lb breaking strength and is commonly supplied in 1 ft increments from a 1,000 ft total coil length. Due to its cut-to-length nature, this item is not returnable.
  • 5/32 in diameter
  • Galvanized steel construction
  • 7 x 19 strand core for flexibility and strength
  • 2,800 Lb breaking strength
  • Available in 1 ft increments from a 1,000 ft total coil length
  • Uncoated surface
Catalog No. 50-005-11905 Supplier MSC Supplier No. 45701521
This galvanized steel wire rope features a 5/32 in diameter and a 7 x 19 strand core. It offers a 2,800 Lb breaking strength and is commonly supplied in 1 ft increments from a 1,000 ft total coil length. Due to its cut-to-length nature, this item is not returnable.
  • 5/32 in diameter
  • Galvanized steel construction
  • 7 x 19 strand core for flexibility and strength
  • 2,800 Lb breaking strength
  • Available in 1 ft increments from a 1,000 ft total coil length
  • Uncoated surface