Learn More
MSC Value Collection Wire Rope Clip, 3/8 in Rope Diameter, Steel, Galvanized
Catalog No. 5000534976
Encompass
This wire rope clip is designed for securing wire rope, featuring a durable steel construction with a galvanized finish for enhanced corrosion resistance. It is suitable for applications requiring a 3/8 inch rope diameter.
  • Clip accessory
  • Designed for 3/8 inch rope diameter
  • Constructed from steel
  • Galvanized finish for corrosion resistance
Catalog No. 50-005-34976 Supplier MSC Supplier No. 53593190
This wire rope clip is designed for securing wire rope, featuring a durable steel construction with a galvanized finish for enhanced corrosion resistance. It is suitable for applications requiring a 3/8 inch rope diameter.
  • Clip accessory
  • Designed for 3/8 inch rope diameter
  • Constructed from steel
  • Galvanized finish for corrosion resistance