Learn More
MSC Value Collection Wire Rope Thimble, 3/4 in Rope Diameter, Steel, Galvanized
Catalog No. 5000528173
Encompass
This wire rope thimble provides crucial protection for the eye of a wire rope, preventing abrasion and crushing, thereby extending the rope's lifespan. Manufactured from durable steel, this thimble is finished with galvanization to offer superior resistance against rust and corrosion, ensuring reliable performance in harsh environments.
  • Designed for 3/4 inch rope diameter.
  • Constructed from steel for robust performance.
  • Features a galvanized finish for corrosion resistance.
Catalog No. 50-005-28173 Supplier MSC Supplier No. 53593117
This wire rope thimble provides crucial protection for the eye of a wire rope, preventing abrasion and crushing, thereby extending the rope's lifespan. Manufactured from durable steel, this thimble is finished with galvanization to offer superior resistance against rust and corrosion, ensuring reliable performance in harsh environments.
  • Designed for 3/4 inch rope diameter.
  • Constructed from steel for robust performance.
  • Features a galvanized finish for corrosion resistance.