Learn More
MSC Value Collection Wire Shelving, 4 Shelves, Starter Unit, 63 in High
Catalog No. 5000516457
Encompass
Quality products at a great value to support your business need.
  • Starter unit for shelving system.
  • 63 in high.
  • 24 in wide.
  • 18 in deep.
  • Black finish.
  • Includes 4 shelves.
  • Supports a load capacity of 3200 lb.
Catalog No. 50-005-16457 Supplier MSC Supplier No. 43382415
Quality products at a great value to support your business need.
  • Starter unit for shelving system.
  • 63 in high.
  • 24 in wide.
  • 18 in deep.
  • Black finish.
  • Includes 4 shelves.
  • Supports a load capacity of 3200 lb.