Learn More
MSC Value Collection Wrap-Around Drum Heater with Thermostat, 4 in Wide, 55 gal, 120V, 1200W
Catalog No. 5000527821
Encompass
This wrap-around drum heater is designed for heating contents within a 55 gallon drum. It operates at 120V and provides 1200W of power, featuring a built-in thermostat that allows for temperature control from 70°F to 425°F.
  • Designed for 55 gal drum capacity.
  • 4 in wide heater band.
  • Operates at 120V.
  • Provides 1200W of heating power.
  • Thermostat controlled temperature range of 70–425°F.
Catalog No. 50-005-27821 Supplier MSC Supplier No. 48650311
This wrap-around drum heater is designed for heating contents within a 55 gallon drum. It operates at 120V and provides 1200W of power, featuring a built-in thermostat that allows for temperature control from 70°F to 425°F.
  • Designed for 55 gal drum capacity.
  • 4 in wide heater band.
  • Operates at 120V.
  • Provides 1200W of heating power.
  • Thermostat controlled temperature range of 70–425°F.