Learn More
MSC Value Collection Zero Setter, 0.00040 in Accuracy, 0.0100 mm Repeatability
Catalog No. 5000528755
Encompass
This zero setter is designed for precise measurement applications. It features an overall height of 50.80 mm, an overall width of 37.00 mm, and an overall length of 37.00 mm. The device offers high accuracy and repeatability for reliable performance.
  • Accuracy of 0.00040 in
  • Repeatability of 0.0100 mm
  • Includes indicator
  • Non-magnetic design
  • Graduation of 0.00050 in
Catalog No. 50-005-28755 Supplier MSC Supplier No. 56451172
This zero setter is designed for precise measurement applications. It features an overall height of 50.80 mm, an overall width of 37.00 mm, and an overall length of 37.00 mm. The device offers high accuracy and repeatability for reliable performance.
  • Accuracy of 0.00040 in
  • Repeatability of 0.0100 mm
  • Includes indicator
  • Non-magnetic design
  • Graduation of 0.00050 in