Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Value Convertible Refrigerator/Freezer PROMO

Catalog No. FBV20FPSS24 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric) Refrigerator:
594 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Refrigerator
FBV20FPSS24 594 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. FBV20FPSS24
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Flexible and economical laboratory storage.

Fisherbrand™ Value Convertible Refrigerator/Freezers offer economical and simple sample storage while also offering increased flexibility of operating as either a refrigerator or a freezer with the push of a button.

Note: Comes configured as a freezer

TRUSTED_SUSTAINABILITY

Specifications

Type General Purpose Refrigerator (Convertible to Freezer)
Capacity (English) Refrigerator 21 cu. ft.
Capacity (Metric) Refrigerator 594 L
Temperature Range (Freezer) -12°C to -24°C
Temperature Range (Refrigerator) 1°C to 12°C
Temperature (Setpoint) Adjustable
Door Style Hinged, Open Left to Right, Non-reversible
No. of Doors 1 (1 compartment)
Shelves 3 Adjustable
Door Shelves 4 fixed shelves
Compressor Bottom-mounted
Defrost Automatic
Operating Modes 2 - Refrigerator or Freezer
Refrigerant R600a
Exterior Finish White-enameled Steel
Insulation Cyclopentane
Interior ABS Plastic
Casters 2 Adjustable Leveling Feet
Color White
Control Type Electronic control
For Use With (Application) Indoor general storage of non-critical samples and reagents
Lighting Interior
Min. Clearance 4 in. on sides, 4 in. at back, 11 in. on top
Typical Lifetime 10 years
Warranty 13 Months Parts and Labor
Dimensions (DxWxH) Exterior 73.5 x 83.2 x 195 cm (28.94 x 32.76 x 76.77 in.)
Dimensions (D x W x H) Interior 57.15 x 67.3 x 175.26 cm (22.5 x 26.5 x 69 in.)
Amperage 2.2 A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115 V, 60 Hz
Frequency 50 Hz
Shipping Weight (English) 244 lb.
Shipping Weight (Metric) 110.6 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.