Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Value Duct Tape

Catalog No. 19072080 Shop All 3M Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-072-080 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-080 Supplier 3M™ Supplier No. 7000049202
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Value Duct Tape is constructed of polyethylene film laminated to cloth with a synthetic rubber adhesive

  • Thin duct tape for light duty applications
  • High tack synthetic rubber adhesive sticks well to many surfaces
  • Easy tear by hand
  • Moisture-resistant polyethylene backing
  • Resists curling and tears off the roll cleanly for simple applications in MRO/construction.
  • Economically priced to best provide a quick solution for simple tasks.
TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Material Synthetic Rubber
Product Type Industrial Tape
For Use With (Application) Design and Construction, Energy, Manufacturing, Mining, Oil and Gas, Transportation
Model 1900
Quantity 24/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.