Learn More
JANT PHARMACAL CORPORATION ACCUSTRIP Value+ Strep A Rapid Test Strip (Throat Swab) 25 Tests/Kit
SDP
Catalog No. NC3457005
Encompass_Preferred
The Accustrip Strep A Value+ Test Strip is a lateral flow, one-step immunoassay for the rapid,qualitative detection of Group A Streptococcal antigen from throat swabs. The test is intended for use as an aid in the diagnosis of Group A Streptococcal infection.
This item is not returnable. View return policy
Catalog No. NC3457005 Supplier JANT PHARMACAL CORPORATION Supplier No. ID513
The Accustrip Strep A Value+ Test Strip is a lateral flow, one-step immunoassay for the rapid,qualitative detection of Group A Streptococcal antigen from throat swabs. The test is intended for use as an aid in the diagnosis of Group A Streptococcal infection.