Learn More
MSC OEM Tools Valve Tappet Feeler Gauge, 30° Offset, 12-Blade, 1 ea.
Catalog No. 5000511655
Encompass
This 12-blade valve tappet feeler gauge is designed for precision clearance measurements in mechanical applications. Featuring an offset 30° angle, it provides access to tight spaces, making it suitable for checking valve lash and other gaps where accuracy is critical.
  • Includes 12 blades constructed from hardened, tempered steel
  • Measurement range from 0.203 mm (0.008 in.) to 0.660 mm (0.026 in.)
  • Blades laser etched with both inches and millimeters
  • Blade length of 10.16 cm (4 in.)
  • Blade protector features a thumb-notch for easy selection and removal
  • Includes a handle
Catalog No. 50-005-11655 Supplier MSC Supplier No. 35340256
This 12-blade valve tappet feeler gauge is designed for precision clearance measurements in mechanical applications. Featuring an offset 30° angle, it provides access to tight spaces, making it suitable for checking valve lash and other gaps where accuracy is critical.
  • Includes 12 blades constructed from hardened, tempered steel
  • Measurement range from 0.203 mm (0.008 in.) to 0.660 mm (0.026 in.)
  • Blades laser etched with both inches and millimeters
  • Blade length of 10.16 cm (4 in.)
  • Blade protector features a thumb-notch for easy selection and removal
  • Includes a handle