Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valutek™ Latex-Free Polypropylene Beard Cover

Catalog No. 191501194
Change view
Click to view available options
Material:
Spunbond Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-150-1194 Spunbond Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-150-1194 Supplier Valutek™ Supplier No. VTBCV18
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured with 100% non-linting, non-woven, spunbond polypropylene fabric to provide comfort and protection. Recommended for Class 10,000 (ISO 7) cleanroom environments.

  • Disposable SPP beard cover
  • Manufactured with 100% non-linting, non-woven, spunbond polypropylene fabric
  • Features a filament covered, unexposed elastic band for secure fit and closure
  • Tested and manufactured in ISO compliant facilities under individual inspection and strict process
  • Recommended for use in a cleanroom Class 10,000+ (ISO 7+) critical environment

Specifications

Type Cleanroom Beard Cover
Material Spunbond Polypropylene
Color White
Disposable Single-use
Latex Free Yes
Certifications/Compliance ISO 7/Class 10,000
For Use With (Application) Full coverage cleanroom contamination control
Length (English) 18 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.