Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valutek™ Half-Finger Nylon Glove Liners

Catalog No. 191501658
Change view
Click to view available options
One Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-150-1658 One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-150-1658 Supplier Valutek™ Supplier No. VTGNLR1/2
This item is not returnable. View return policy

Liners are constructed of lightweight, non-linting 100% continuous filament virgin nylon. Seamless design limits skin irritation caused by direct contact with latex, nitrile, or PVC gloves.

  • 100% knitted nylon material is lightweight and non-linting
  • Fits comfortably under latex, nylon, or PVC gloves
  • Seamless design limits skin irritation caused by direct contact with glove material
  • Tested and manufactured in ISO-compliant facilities
  • One size fits most
  • Ambidextrous

Specifications

Coating Material Uncoated
Color White
Hand Ambidextrous
Laboratory/Cleanroom Use Yes
Material Nylon
Size Universal
Finger Style Half
For Use With (Application) Cleanroom Class 100-1000 (ISO 5-6) Critical Environments
Glove Cuff Style Knit
Series VTGNLR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.