Learn More
JANT PHARMACAL CORPORATION Accutest VALUE+™ hCG Urine/Serum Pregnancy Test Cassette (CLIA-Waived for urine)
SDP
Catalog No. NC3456995
Encompass_Preferred
Rapid test for the detection of human chorionic gonadotropin (hCG) in serum and urine specimens. This test is used to obtain a visual, qualitative result and is intended for professional and laboratory use.
Catalog No. NC3456995 Supplier JANT PHARMACAL CORPORATION Supplier No. PF864
Rapid test for the detection of human chorionic gonadotropin (hCG) in serum and urine specimens. This test is used to obtain a visual, qualitative result and is intended for professional and laboratory use.