Learn More
JANT PHARMACAL CORPORATION Accutest VALUPAK hCG Urine Pregnancy Test Cassette (CLIA Waived) 40 tests/kit box
SDP
Catalog No. NC3457003
Encompass_Preferred
Introducing our new Accutest ValuPak™ Pregnancy Test kit for the early detection of pregnancy. You get the same quality, more value and convenience for less. The Accutest ValuPak™ Pregnancy Test is a rapid test for the detection of human chorionic gonadotropin (hCG) in urine specimens, used to obtain a visual, qualitative result and is intended for professional and laboratory use.
This item is not returnable. View return policy
Catalog No. NC3457003 Supplier JANT PHARMACAL CORPORATION Supplier No. PF453
Introducing our new Accutest ValuPak™ Pregnancy Test kit for the early detection of pregnancy. You get the same quality, more value and convenience for less. The Accutest ValuPak™ Pregnancy Test is a rapid test for the detection of human chorionic gonadotropin (hCG) in urine specimens, used to obtain a visual, qualitative result and is intended for professional and laboratory use.