Learn More
KIMBLE™ ValueWare™ Erlenmeyer Flask, Narrow Mouth
SDP
Catalog No. 01672730 Shop All DWK Life Sciences Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
01-672-730 Autoclavable
1 options
Catalog No. 01-672-730 Supplier KIMBLE™ Supplier No. 5650050
Erlenmeyer Flask made of borosilicate glass 3.3.
  • Complies with ASTM Specification E1404, Type I, Class 1 requirements
  • Manufactured from borosilicate glass 3.3
  • Tops are reinforced and tooled with a rounded finish, containing more glass for maximum mechanical strength
  • Body has sturdy walls and long tapered outside contour to minimize chipping when struck or rubbed together
  • All flasks have durable, white, ceramic enamel scales indicating approximate volumes at various levels
  • Useful in measuring and mixing solutions where a high degree of accuracy is unnecessary
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL
Material Borosilicate Glass (3.3)
Product Type Erlenmeyer Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Round
Surface Treatment None