Learn More
DWK Life Sciences Kimble™ ValueWare™ Volumetric Flasks, Class A
SDPSCI_ED
Catalog No. S00128 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S00128 2000 mL
S00120 10 mL
S00121 25 mL
S00122 50 mL
S00123 100 mL
S00125 250 mL
S00124 200 mL
S00126 500 mL
S00127 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. S00128 Supplier DWK Life Sciences Supplier No. 580142000
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Calibrated To Contain.
  • Kimax™ KG-33 glass Single graduation ring blasted on the neck
  • With plug stopcock ground glass stopper and marking spot
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E288, Class A unserialized requirements.
TRUSTED_SUSTAINABILITY
Specifications
Calibration Calibrated To Contain
Closure Material Glass
Capacity (Metric) 2000 mL
Material Glass
Closure Size 27