Learn More
International Enviroguard™ ValuGuard MP™ Lightweight Microporous Coveralls with Elastic Wrists and Open Ankles
Catalog No. 17184060 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Large
Catalog No. Size
17-184-060 Large
1 options
Catalog No. 17-184-060 Supplier International Enviroguard™ Supplier No. 8112L
Add to Cart
Add to Cart
Economical coveralls are made of lightweight microporous fabric that provides high moisture vapor transmission rate (HMVT) to help workers stay cool and prevent heat stress.
  • Coveralls protect against particulates, light liquid splashes, and sprays in nonhazardous work environments
  • Disposable coveralls provide a basic barrier against varnish, resins, fiberglass, and paints
  • Keep productivity high and risk level low when wearing ValuGuard MP coveralls
  • Secure, elastic wrists help keep sleeves in place to eliminate the possibility of skin exposure
  • Standard zipper and set-in sleeves
  • Environmentally friendly; can be recycled after use
Specifications
Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
Waist Style Standard
Ankle Style Open
Cuff Style Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Liquid, Particle, Tears
Latex Free Yes
Model 8112
No. of Pockets None