Learn More
ValueMax™ Easy-Breathe™ Lab Jackets - Medical Blue
Catalog No. 23900714
Change view
Click to view available options
Size:
4X-Large
5X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
23-900-714 4X-Large
23-900-715 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-900-714 Supplier ValueMax™ Supplier No. 3530MB4XL
This item is not returnable. View return policy
Affordable apparels combined with fluid resistance and breathability
  • Silky soft “SMS” fabrics
  • Anti-static treated and no clinging
  • Snap front, three pockets
  • Knit cuff and collar
  • Hip length and bright colors for any operations
  • Meet AAMI level 2 fluid resistant standard
Specifications
Size 4X-Large
Color Medical Blue
Collar Type Knit
Cuff Style Knit