Learn More
ValueMax™ Extra-Safe™ Lab Coats —
Catalog No. 23900519D
Change view
Click to view available options
Collar Type:
Knit
Size:
Small
Medium
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Collar Type Size
23-900-519D Knit X-Large
23-900-519B Knit Medium
23-900-519A Knit Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-900-519D Supplier ValueMax™ Supplier No. 3660WHXLK
This item is not returnable. View return policy
Static-free — no clinging
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Color: White
  • Fluid-resistant, breathable
  • Snap front, three pockets, knit cuffs
  • Available with knit or traditional collar
  • Knee-length
  • Autoclavable and washable up to five times
  • Latex-free, fire retardant
Specifications
Product Type General Purpose Lab Coat
Size X-Large
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Disposable Single-use
Style Knee-length
No. of Pockets 3
Collar Type Knit
Latex Free Yes
Model 3660WH
Thickness 3-ply
Construction 3-ply SMS