Learn More
ValueMax™ Extra-Safe™ Lab Coats — Blueberry
Catalog No. 23900522C
Change view
Click to view available options
Size:
Large
Catalog No. Size
23-900-522C Large
1 options
This item is not returnable. View return policy
Catalog No. 23-900-522C Supplier ValueMax™ Supplier No. 3660BBL
This item is not returnable. View return policy
Static-free — no clinging
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid-resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Knee length
  • Autoclavable and washable up to five times
  • Latex-free, fire retardant
Specifications
Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material SMS
Color Blueberry
Closure Type Snaps
Disposable Single-use
Style Knee-length
No. of Pockets 3
Latex Free Yes
Model 3660BB
Thickness 3-ply
Construction 3-ply SMS