Learn More
ValueMax™ Extra-Safe™ Lab Coats — Ceil Blue
Catalog No. 23900513G
Change view
Click to view available options
Size:
Small
4X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
23-900-513G 4X-Large
23-900-513A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-900-513G Supplier ValueMax™ Supplier No. 3660CB4XL
This item is not returnable. View return policy
Static-free — no clinging
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid-resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Knee length
  • Autoclavable and washable up to five times
  • Latex-free, fire retardant
Specifications
Size 4X-Large
Material SMS
Color Ceil Blue
Closure Type Snaps
Disposable Single-use
Style Knee-length
No. of Pockets 3
Latex Free Yes
Thickness 3-ply
Construction 3-ply SMS