Learn More
ValueMax™ Extra-Safe™ Lab Jackets — Ceil Blue
Catalog No. 23900502I
Change view
Click to view available options
Size:
Large
X-Large
5X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
23-900-502I 5X-Large
23-900-502E X-Large
23-900-502D Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-900-502I Supplier ValueMax™ Supplier No. 3630CB5XL
This item is not returnable. View return policy
Static free — no clinging
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Hip length
  • Autoclavable and washable up to five times
  • Latex free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Size 5X-Large
Material Polypropylene
Color Ceil Blue