Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valutek™ Polypropylene Cleanroom Sleeves

Catalog No. 191500828
Change view
Click to view available options
Length (English):
21 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-150-0828 21 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-150-0828 Supplier Valutek™ Supplier No. VTSLVC21
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Disposable polypropylene sleeves are lightweight, strong, and durable. Recommended for use in a cleanroom Class 100-1,000 (ISO 5-6) critical environments.

  • Nonwoven 100 percent spun-bond polypropylene
  • Comfortable, non-latex elastic cuffs at wrist and arm
  • Length: 53.34 cm (21 in.)

Compliance

Product conforms to the following test standards: IES-RP-CC-003.3

Specifications

Product Type Controlled Environment Sleeve
Material Polypropylene
Sterility Non-sterile
Disposable Single-use
Cuff Style Elastic
Latex Free Yes
Model VTSLVC-21
Length (English) 21 in.
Length (Metric) 53.3 cm
Cleanroom Class Class 100-1000 (ISO 5-6)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.