Learn More
Chemglass Life Sciences Valve, 0-12mm, 90°, PTFE Tip
SDP
Catalog No. 501532032
Encompass_Preferred
Valve, 0-12mm, 90, PTFE Tip. Hi-vac valve with two sidearms sealed to the barrel at a 90 angle to one another. Designed with a precision external thread for fine control.
Catalog No. 50-153-2032 Supplier Chemglass Life Sciences Supplier No. CGK55103
Valve, 0-12mm, 90, PTFE Tip. Hi-vac valve with two sidearms sealed to the barrel at a 90 angle to one another. Designed with a precision external thread for fine control.