Learn More
Chemglass Life Sciences Valve, 0-12mm, 180°, PTFE Tip
SDP
Catalog No. 501532035
Encompass_Preferred
Valve, 0-12mm, 180, PTFE Tip. Hi-vac valve with two sidearms sealed to the barrel at a 180 angle to one another and on the same plane.
Catalog No. 50-153-2035 Supplier Chemglass Life Sciences Supplier No. CGK55303
Valve, 0-12mm, 180, PTFE Tip. Hi-vac valve with two sidearms sealed to the barrel at a 180 angle to one another and on the same plane.