Learn More
Chemglass Life Sciences Valve, 0-4mm, 1-Arm, PTFE Tip
SDP
Catalog No. 501532036
Encompass_Preferred
Valve, 0-4mm, 1-Arm, PTFE Tip. This hi-vac valve has a single sidearm seal at a 90 angle to the barrel. This inexpensive configuration allows for the lower portion of the barrel to be sealed directly to the apparatus being constructed, making for more compact, stronger assemblies. Designed with a precision external thread for fine control.
Catalog No. 50-153-2036 Supplier Chemglass Life Sciences Supplier No. CGK56001
Valve, 0-4mm, 1-Arm, PTFE Tip. This hi-vac valve has a single sidearm seal at a 90 angle to the barrel. This inexpensive configuration allows for the lower portion of the barrel to be sealed directly to the apparatus being constructed, making for more compact, stronger assemblies. Designed with a precision external thread for fine control.