Learn More
Chemglass Life Sciences Valve, 0-4mm, 90°, Exposed O-ring Tip
SDP
Catalog No. 501532019
Encompass_Preferred
Valve, 0-4mm, 90, Exposed O-ring Tip. Hi-vac valve with two sidearms sealed to the barrel at a 90 angle to one another. Designed with a precision external thread for fine control.
Catalog No. 50-153-2019 Supplier Chemglass Life Sciences Supplier No. CGK52501
Valve, 0-4mm, 90, Exposed O-ring Tip. Hi-vac valve with two sidearms sealed to the barrel at a 90 angle to one another. Designed with a precision external thread for fine control.