Learn More
Chemglass Life Sciences Valve, 0-4, In-Line, Exposed O-Ring Tip
SDP
Catalog No. 501532024
Encompass_Preferred
Valve, 0-4, In-Line, Exposed O-Ring Tip. Hi-vac valve with bent arms to minimize hold up of fluids that would occur in conventional valves. Designed with a precision external thread for fine control.
Catalog No. 50-153-2024 Supplier Chemglass Life Sciences Supplier No. CGK52901
Valve, 0-4, In-Line, Exposed O-Ring Tip. Hi-vac valve with bent arms to minimize hold up of fluids that would occur in conventional valves. Designed with a precision external thread for fine control.