Learn More
Chemglass Life Sciences Valve, 0-8mm, Hi-Vac, 1-Arm, Exposed O-ring
SDP
Catalog No. 501532026
Encompass_Preferred
Valve, 0-8mm, Hi-Vac, 1-Arm, Exposed O-ring. This style hi-vac valve has a single sidearm seal at a 90 angle to the barrel. This inexpensive configuration allows for the lower portion of the barrel to be sealed directly to the apparatus being constructed, making for more compact, stronger assemblies. Designed with a precision external thread for fine control.
Catalog No. 50-153-2026 Supplier Chemglass Life Sciences Supplier No. CGK53402
Valve, 0-8mm, Hi-Vac, 1-Arm, Exposed O-ring. This style hi-vac valve has a single sidearm seal at a 90 angle to the barrel. This inexpensive configuration allows for the lower portion of the barrel to be sealed directly to the apparatus being constructed, making for more compact, stronger assemblies. Designed with a precision external thread for fine control.