Learn More
Chemglass Life Sciences Valve, 0-8mm, 90°, Exposed O-ring Tip
SDP
Catalog No. 501532020
Encompass_Preferred
Valve, 0-8mm, 90, Exposed O-ring Tip. Hi-vac valve with two sidearms sealed to the barrel at a 90 angle to one another. Designed with a precision external thread for fine control.
Catalog No. 50-153-2020 Supplier Chemglass Life Sciences Supplier No. CGK52502
Valve, 0-8mm, 90, Exposed O-ring Tip. Hi-vac valve with two sidearms sealed to the barrel at a 90 angle to one another. Designed with a precision external thread for fine control.