Learn More
Chemglass Life Sciences Valve, 0-8mm, 180°, PTFE Tip
SDP
Catalog No. 501532034
Encompass_Preferred
Valve, 0-8mm, 180, PTFE Tip. Hi-vac valve with two sidearms sealed to the barrel at a 180 angle to one another and on the same plane.
Catalog No. 50-153-2034 Supplier Chemglass Life Sciences Supplier No. CGK55302
Valve, 0-8mm, 180, PTFE Tip. Hi-vac valve with two sidearms sealed to the barrel at a 180 angle to one another and on the same plane.