Learn More
Grainger Schneider Electric Hydronic Globe Valve, Bronze, 1-1/4 in., 20 Cv, 1 ea
Catalog No. 502761066
Encompass_Preferred
This hydronic globe valve is designed for precise control of chilled water, hot water, or steam applications within HVAC systems. Constructed with a durable bronze body, this 2-way valve features a stem-up to close action and is compatible with various electric or pneumatic actuators for versatile installation. It is built to ensure reliable performance with a self-adjusting packing design and high-quality materials.
  • Bronze body, plug, and seat construction
  • Stainless steel stem with spring-loaded TFE packing
  • 20 Cv flow coefficient
  • ANSI Class IV leakage rating of 0.01% of Cv
  • Operating temperature range of 20°F to 281°F
  • 1-1/4 in. NPT connection
  • Media compatibility includes hot or chilled water, glycol up to 50%, and steam up to 35 psig
Catalog No. 50-276-1066 Supplier Grainger Supplier No. 161Z07
This hydronic globe valve is designed for precise control of chilled water, hot water, or steam applications within HVAC systems. Constructed with a durable bronze body, this 2-way valve features a stem-up to close action and is compatible with various electric or pneumatic actuators for versatile installation. It is built to ensure reliable performance with a self-adjusting packing design and high-quality materials.
  • Bronze body, plug, and seat construction
  • Stainless steel stem with spring-loaded TFE packing
  • 20 Cv flow coefficient
  • ANSI Class IV leakage rating of 0.01% of Cv
  • Operating temperature range of 20°F to 281°F
  • 1-1/4 in. NPT connection
  • Media compatibility includes hot or chilled water, glycol up to 50%, and steam up to 35 psig