Learn More
Alta Robbins VALVE 1/4 NPT X 1/4
SDP
Catalog No. NC9964125
Encompass
Valve, 1/4 NPT x 1/4, 1.94 in. Operating pressure vacuum 3500psi, Safety factor: 4: 1, Cv: 0.31, Leakage: Zero, Orifice dia. 0.187 in., Body Material 303 Stainless Steel, Bonnet Material 303 Stainless Steel, Stem Material 303 SS
Catalog No. NC9964125 Supplier Alta Robbins Supplier No. 3141
Valve, 1/4 NPT x 1/4, 1.94 in. Operating pressure vacuum 3500psi, Safety factor: 4: 1, Cv: 0.31, Leakage: Zero, Orifice dia. 0.187 in., Body Material 303 Stainless Steel, Bonnet Material 303 Stainless Steel, Stem Material 303 SS