Learn More
Waters Corp Valve, 1/8 in. Compression, 1500 psi, Ball Type
Catalog No. 502392367 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This high-pressure ball valve is designed with 1/8-inch compression fittings and can withstand pressures up to 1500 psi. It is suitable for fluid control in HPLC and other analytical equipment, offering stable flow control and durability, and can be used as a spare part for maintenance.
  • Designed with 1/8-inch compression fittings
  • Withstands pressures up to 1500 psi
  • Suitable for fluid control in HPLC and other analytical equipment
  • Offers stable flow control and durability
  • Can be used as a spare part for maintenance
  • Functions as a spare parts kit
Catalog No. 50-239-2367 Supplier Waters Corp Supplier No. 700007291
This high-pressure ball valve is designed with 1/8-inch compression fittings and can withstand pressures up to 1500 psi. It is suitable for fluid control in HPLC and other analytical equipment, offering stable flow control and durability, and can be used as a spare part for maintenance.
  • Designed with 1/8-inch compression fittings
  • Withstands pressures up to 1500 psi
  • Suitable for fluid control in HPLC and other analytical equipment
  • Offers stable flow control and durability
  • Can be used as a spare part for maintenance
  • Functions as a spare parts kit